Skip to Content
Merck

Skip To

1.50252

Purospher® STAR RP-18 endcapped (5 µm) HPLC Columns

L × I.D. 250 mm x 4 mm LiChroCART HPLC cartridge, manuCART holder required for use

Synonym(s):

Purospher® STAR RP-18 endcapped (5 µm) LiChroCART 250-4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52
Particle size:
5 μm
Matrix active group:
C18 (RP18), C18 bonding phase
Pore size:
120 Å pore size
Matrix:
fully porous particle
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Purospher® STAR RP-18 endcapped (5 µm), L × I.D. 250 mm x 4 mm LiChroCART HPLC cartridge, manuCART holder required for use

material

stainless steel column

Quality Segment

agency

suitable for USP L1

product line

Purospher®

feature

endcapped

packaging

1 ea of

extent of labeling

17% Carbon loading

parameter

250 bar pressure, 65 °C max. temp.

technique(s)

HPLC: suitable, LC/MS: suitable, mass spectrometry (MS): suitable (High)

column L × I.D.

25 cm × 4 mm

surface area

330 m2/g

matrix

fully porous particle

matrix active group

C18 (RP18), C18 bonding phase

particle size

5 μm

pore size

120 Å pore size

operating pH range

1.5-10.5

separation technique

reversed phase

storage temp.

no temp limit

General description

Purospher® STAR RP-18 endcapped, 5 µm HPLC columns are designed for universal use and allow the separation of basic, neutral, and metal chelating compounds with simple mobile phases and excellent peak symmetry. They are based on ultra-pure fully porous silica particles (Type B). These columns offer an outstanding stability from pH 1.5 to 10.5 over a wide temperature range, and suitability for up to 100% aqueous mobile phases. Plus, they demonstrate best all-round retention characteristics, as proven by the Tanaka test. Thanks to these features, Purospher® STAR RP-18 endcapped, 5 µm HPLC columns allow maximum flexibility to use the best chromatographic conditions for any separation including reversed phase.Purospher® STAR RP-18 endcapped, 5 µm LiChroCART 250-4 is a cartridge column which requires a manuCART cartridge column holder part number 1.51486.0001. The manuCART can be used to hold one cartridge column with or without direct integration of a 4-4 mm guard column. The recommended guard column is Purospher® STAR RP-18 endcapped (5 µm) LiChroCART 4-4 part number 1.50250.0001. LiChroCART cartridge columns mounted with a manuCART cartridge column holder are hand-tight only and have therefore a maximum pressure stability of 250 bar. For higher pressure stability requirements Purospher® STAR RP-18 endcapped, 5 µm Hibar® RT 250-4 HPLC columns are available part number 1.150037.0001.The column dimension of 250 mm length x 4 mm I.D. is the optimal choice for high resolution HPLC separation of complex samples.Item 1.50252.1003 is a Validation kit containing 3 Purospher® STAR RP-18 endcapped, 5 µm LiChroCART 250-4 cartridge columns from 3 different base material lots.Learn more about Purospher® STAR HPLC and UHPLC ColumnsLearn more about the Supelco HPLC Column portfolio and Method development:Brochure: HPLC and UHPLC Column Selection GuideBrochure: A Practical Guide to HPLC - Method Development Guide

Application

Intended for the separation of aflatoxins, aldehydes, alkaloids, antibiotics, catecholamines, explosives, oligonucleotides, esters, fat-soluble vitamins, lipids, fatty acids, flavonoids, ketones, nitrosamines, nucleosides, PAHs, PCBs, peptides, pesticides, phenols, phthalates, preservatives, steroids, metabolized steroids, sulfonamides, sweeteners, and water-soluble vitamins.

Features and Benefits

  • Good suitability for LC-MS applications

Analysis Note

Pressure: ≤ 130 bar
Theoretical Plates (N/m) (Toluene): ≥ 80000
Symmetry (Toluene): 0.8 - 1.4
Capacity factor (Toluene): ≥ 1.5

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANUCART is a trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
150250150254150241
description

L × I.D. 250 mm x 4 mm LiChroCART HPLC cartridge, manuCART holder required for use

description

L × I.D. 4 mm x 4 mm LiChroCART Guard-cartridge

description

L × I.D. 250 mm x 3 mm LiChroCART HPLC cartridge, manuCART holder required for use

description

L × I.D. 55 mm x 2 mm LiChroCART HPLC cartridge, manuCART holder required for use

matrix active group

C18 (RP18), C18 bonding phase

matrix active group

C18 (RP18), C18 bonding phase

matrix active group

C18 (RP18), C18 bonding phase

matrix active group

C18 (RP18), C18 bonding phase

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

agency

suitable for USP L1

agency

suitable for USP L1

agency

suitable for USP L1

agency

suitable for USP L1

pore size

120 Å pore size

pore size

120 Å pore size

pore size

120 Å pore size

pore size

120 Å pore size

product line

Purospher®

product line

Purospher®

product line

Purospher®

product line

Purospher®

matrix

fully porous particle

matrix

fully porous particle

matrix

fully porous particle

matrix

fully porous particle


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters