Skip to Content
Merck
All Photos(1)

Key Documents

1.44018

Millipore

Hycon® Contact Slides

Rose-Bengal Chloramphenicol Agar, for yeasts, for molds, aseptically filled, single packed of 20 units, suitable for surface monitoring

Synonym(s):

Hycon® Contact Slides YM, Rose-Bengal Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Quality Level

sterility

aseptically filled

form

plate (ready-to-use)

feature

ready-to-use

packaging

single packed of 20 units

manufacturer/tradename

HYCON®

pH

7.1 ( in H2O)

mp

>95 °C

application(s)

cosmetics
food and beverages
pharmaceutical

surface monitoring

compatibility

stand alone

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Contact Slides YM are developed for easy and flexible microbiological monitoring of flat and curved surfaces. Each contact slide is individually sealed in the transparent and flexible primary backing film made from PET and provides a rectangular surface of 25 cm2. Rose-Bengal Chloramphenicol Agar is a selective medium for the enumeration of yeasts and molds. Chloramphenicol, streptomycin and Rose-Bengal are included as selective agents to inhibit bacterial growth. Rose-Bengal also restricts the size and height of colonies of the rapidly growing molds, thereby facilitates the detection of slow-growing fungi by preventing overgrowth.

Application

Contact Slides YM are designed for the determination of the yeast and mold count on dry, sanitized surfaces as well as personnel in controlled clean room environments of production lines, i.e. food and beverages as well as non-sterile and terminally sterilized pharmaceuticals.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

For easy and flexible microbiological monitoring of surfaces in the food & beverage industry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service