1.40000
Extran® MA 05
liquid, alkaline, phosphate-free concentrate
About This Item
Recommended Products
Quality Level
product line
Extran®
form
liquid
greener alternative product characteristics
Design for Degradation
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
impurities
≤0.002% Phosphate (PO4)
1.5-2.5% Free alkali (as NaOH)
8.5-10% sequestering agent content (alkalimetric)
pH
13 (20 °C in H2O)
solubility
water: soluble
density
1.06 g/cm3 at 20 °C
anion traces
phosphate (PO43-): ≤0.002%
application(s)
cleaning products
greener alternative category
storage temp.
15-25°C
General description
Linkage
Analysis Note
Density (d 20 °C/ 4 °C): 1.05 - 1.09
Free alkali (as NaOH): 1.5 - 2.5 %
Phosphate (PO₄): ≤ 0.002 %
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service