Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352202
NACRES:
NA.42
eCl@ss:
32160405
Pricing and availability is not currently available.
Quality Segment
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
NCBI accession no.
UniProt accession no.
shipped in
wet ice
Biochem/physiol Actions
Protein Target: PKA
Target Sub-Family: AGC
Physical form
Lyophilized powder
Preparation Note
Lyophilized: 2 years at 4°C. Rehydrated: 1 year at -20°C.
Analysis Note
Routinely evaluated by using PKA catalytic subunit (14-114) to phosphorylate the substrate.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| description PKA Substrate Peptide primarily used in Kinase Assays. | description PKC Substrate Peptide primarily used in Kinase Assays. | description Akt/SGK Substrate Peptide primarily used in Kinase Assays. | description MAPKAP Kinase Substrate Peptide primarily used in Kinase Assays. |
| technique(s) activity assay: suitable (kinase) | technique(s) activity assay: suitable (kinase) | technique(s) activity assay: suitable (kinase) | technique(s) activity assay: suitable (kinase) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 300 |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| shipped in wet ice | shipped in wet ice | shipped in dry ice | shipped in wet ice |
| manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® | manufacturer/tradename Upstate® |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



