Skip to Content
Merck

1.19899

Nitrite standard solution

traceable to SRM from NIST NaNO2 in H2O 1000 mg/l NO2 Certipur®

Synonym(s):

Nitrite standard for aqueous matrices ready foruse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

technique(s)

ion chromatography: suitable

pH

7 (20 °C in H2O)

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

Analysis Note

Concentration     β (NO₂⁻): 990 - 1010 mg/l
Determination method: Cerimetric titration.
The content of this solution was determined by back-titration with Ammonium(II) ferrous sulfate (article number 1.03792) standardized by Cerium(IV) sulfate (article number 1.09092) against volumetric standard Iron(II)ethylendiammonium sulfate (article number 1.02402). The expanded measurement uncertainty is ± 0.7 % (k=2 coverage factor for 95% coverage probability). The certified value is traceable to primary standard NIST SRM 8040 (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard Iron(II)ethylendiammonium sulfate measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.

Other Notes

We now offer a certified reference material alternative for this product that has the same composition and is manufactured under ISO 17034 accreditation.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Nitrite standard solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library