1.15974
Elastica van Gieson staining kit
suitable for histology, for connective tissue
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116124
NACRES:
NA.41
Recommended Products
Quality Level
IVD
for in vitro diagnostic use
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
2-8°C
General description
Elastica van Gieson staining kit - for connective tissue is a ready to use staining kit used for human-medical cell diagnosis and the method is principally used to visualize elastic fibers in tissue sections. in for example, histological sections of e. g. the liver, the kidney, the intestine, the placenta and similar.
Various staining methods are used to demonstrate elastic fibers in tissue sections. Brilliant staining results are usually achieved when using the resorcinol-fuchsin solution according to Weigert in combination with a picrofuchsin solution acc. to van Gieson, (Product number 1.00199) and nuclear staining with Weigert′s iron hematoxylin solution (Product number 1.15973). These solutions have now been combined in the Elastica van Gieson kit for the detection of elastic fibers in histological sections. The kit contains the set of four ready-to-use solutions and will result in the visualization of black-brown nuclei, black-stained elastic fibers, red collagen and yellow-stained muscles.
The package is sufficient for 400 - 500 applications.
The Elastica van Gieson kit is an IVD & CE product and can be used in the clinical laboratory for diagnostics. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Various staining methods are used to demonstrate elastic fibers in tissue sections. Brilliant staining results are usually achieved when using the resorcinol-fuchsin solution according to Weigert in combination with a picrofuchsin solution acc. to van Gieson, (Product number 1.00199) and nuclear staining with Weigert′s iron hematoxylin solution (Product number 1.15973). These solutions have now been combined in the Elastica van Gieson kit for the detection of elastic fibers in histological sections. The kit contains the set of four ready-to-use solutions and will result in the visualization of black-brown nuclei, black-stained elastic fibers, red collagen and yellow-stained muscles.
The package is sufficient for 400 - 500 applications.
The Elastica van Gieson kit is an IVD & CE product and can be used in the clinical laboratory for diagnostics. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Sens. 1 - STOT SE 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Anna Filip et al.
Frontiers in cardiovascular medicine, 9, 848680-848680 (2022-10-01)
Amine oxidase copper containing 3 (AOC3) displays adhesion between leukocytes and endothelial cells and enzymatic functions. Given its controversial role in atherogenesis, we proposed to investigate the involvement of AOC3 in the formation of atherosclerotic plaques in ApoE-/-AOC3-/- mice and
Holger Cynis et al.
EMBO molecular medicine, 3(9), 545-558 (2011-07-21)
Acute and chronic inflammatory disorders are characterized by detrimental cytokine and chemokine expression. Frequently, the chemotactic activity of cytokines depends on a modified N-terminus of the polypeptide. Among those, the N-terminus of monocyte chemoattractant protein 1 (CCL2 and MCP-1) is
Gunther Felmerer et al.
The Journal of surgical research, 253, 294-303 (2020-05-15)
Lipedema is a common adipose tissue disorder affecting women, characterized by a symmetric subcutaneous adipose tissue deposition, particularly of the lower extremities. Lipedema is usually underdiagnosed, thus remaining an undertreated disease. Importantly, no histopathologic or molecular hallmarks exist to clearly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service