Skip to Content

1.12073

Boron nitride

Synonym(s):

Boron nitride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82
UNSPSC Code:
12352301
EC Index Number:
233-136-6
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

potency

>2000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

pH

5-8 (20 °C, 100 g/L in H2O, slurry)

mp

2700-3000 °C

density

2.34 g/cm3 at 20 °C

bulk density

120 kg/m3

storage temp.

no temp limit

SMILES string

B#N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Application

  • Biocompatibility and Biomedical Applications: Explores the biocompatibility of boron nitride nanomaterials and their potential applications in the biomedical field, focusing on drug delivery systems and bioimaging (Merlo et al., 2018).
  • Boron Nitride Nanosheets for Polymer Composites: Discusses the incorporation of boron nitride nanosheets into polymer matrices to enhance mechanical strength, thermal stability, and electrical insulation properties of composites (Rasul et al., 2021).

Analysis Note

Identity: passes test


Still not finding the right product?

Explore all of our products under Boron nitride


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library