Skip to Content

1.11499

Geobacillus stearothermophilus Spore Suspension

For detection antimicrobial residues, suitable for microbiology

Synonym(s):

G. stearothermophilus, Biological indicators

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
41116130
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

EP, USP

form

liquid (ready-to-use)

feature

ready-to-use

application(s)

food and beverages, microbiology

storage temp.

2-8°C

suitability

Bacillus geobacillus

General description

Detection of antimicrobial residues in meat and other foodstuffs of animal origin is based on agar diffusion, using spores of Geobacillus stearothermophilus ATCC 7953 (sporulation optimized) as test organisms. Antibiotic and sulfonamide residues inhibit the growth of the test organism, which is indicated by the formation of inhibition zones. These inhibition zones remain purple in color whilst the rest of the nutrient medium turns yellow. Cleaning agents, disinfectants and preservatives do not influence the test.

Application

The Geobacillus stearothermophilus spore suspension is used in a routine qualitative procedure to detect antimicrobial residues such as antibiotics, sulfonamides and other chemotherapeutics in meat and other foodstuffs of animal origin. It is used in conjunction with the test agar for performing the antibiotics sulfonamide residue test according to KUNDRAT (1.10662).

Analysis Note

Growth (after 3 to 3.5 hours / 65 ° C): passes test
Colour change to yellow (after 3 to 3.5 hours / 65 ° C): conforms
Growth test (Test agar for the residue test acc. to KUNDRAT): passes test
Growth test (1 week/refrigerator): passes test
Growth promotion test performed with the following antibiotic testplates:
Gentamicin 10 µg; Gentamicin 30 µg; Penicilin 10 IE; Streptomicin 10 µg;
Incubation: 3 - 3.5 hrs. 65 °C.
Reference: Test agar for the residue test acc. to KUNDRAT.
Number of viable spores G. stearothermophilus ATCC 7953: 5E+07 - 5E+08 CFU/ml
Incubation: 24 - 48 hrs. 65 °C aerobic.
Reference: Tryptic soy agar.
Microbiological purity: passes test
Microbiological purity verified with Blood-agar and Sabouraud-4% dextrose agar with chloramphenicol.
Incubation: minimum 72 hrs. 35 °C aerobic.


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library