Skip to Content

1.11138

Phosphate Test Kit, colorimetric

1.0-10 mg/L (P₂O₅), 1.3-13.4 mg/L (PO₄³⁻), MQuant®

Synonym(s):

Phosphate Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate Test, colorimetric, 1.0-10 mg/L (P2O5), 1.3-13.4 mg/L (PO43-), for use with MCOLORTEST®

specific analyte(s)

phosphate

Quality Segment

measuring range

1.0-10 mg/L (P2O5), 1.3-13.4 mg/L (PO43-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, PMB, with color-matching vessel 1.0 - 2.5 - 5.0 - 7.5 - 10 mg/l P2O5 1.3 - 3.3 - 6.7 - 10.0 - 13.4 mg/l PO43- MQuant®
This MQuant® Phosphate test (formally MColortest®) measures orthophosphate in:
  • Groundwater, surface water, and seawater
  • Waters from aquaculture
  • Drinking water and mineral water
  • Industrial water, prozess water, and wastewater
  • Electroplating wastewater
  • Boiler water and boiler feed water
  • Food after appropriate sample pretreatment
  • Soils and fertilizers after appropriate sample pretreatment

Method
This test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation. The color matching vessel is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments. In sulfuric solution orthophosphate ions (PO43-) react with molybdate ions to form molybdophosphoric acid. This is reduced to phosphomolybdenum blue (PMB). The phosphate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color zones of a color matching vessel.
Using the refill pack 1.08046 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Application


  • Zinc and Silicon Nano-Fertilizers Influence Ionomic and Metabolite Profiles in Maize to Overcome Salt Stress: This research utilizes phosphate tests in the study of plant physiology, specifically to gauge the efficacy of nano-fertilizers in enhancing phosphate uptake under salt stress conditions, providing insights into sustainable agricultural practices (Shoukat A et al., 2024).
  • Investigation of Mechanical Properties of Polymer-Infiltrated Tetrapodal Zinc Oxide in Different Variants: Applies phosphate testing in the field of materials science to assess the interaction of phosphate with novel composite materials, thereby informing the development of more durable and efficient materials for various industrial applications (Scherer F et al., 2024).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Phosphate Test


pictograms

CorrosionExclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library