Skip to Content
Merck

1.09928

Potassium dichromate solution

c(K₂Cr₂O₇) = 1/₆₀ mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®

Synonym(s):

K₂Cr₂O₇ Calibration Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352302
NACRES:
NA.21
Grade:
reagent grade
Form:
liquid
Solubility:
water: soluble
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Potassium dichromate solution, for 1000 ml, c(K2Cr2O7) = 1/60 mol/l (0.1 N) Titrisol®

Quality Segment

grade

reagent grade

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Redox Reactions

concentration

1/60 M

technique(s)

titration: suitable

pH

4.1 (20 °C in H2O)

solubility

water: soluble

density

1.06 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Facile synthesis of heterojunction of MXenes/TiO2 nanoparticles: This research involves the use of potassium dichromate solution in the synthesis of advanced nanomaterials for environmental remediation, particularly for the removal of hexavalent chromium from water, underscoring its significance in environmental science and nanotechnology (Wang H et al., 2020).


Analysis Note

Concentration after dilution to 1 liter: c(K₂Cr₂O₇) = 1/60 mol/l
Amount-of-substance concentration: 1/60 mol/l
The concentration of this solution was determined with sodium thiosulfate standard solution (article number 1.09147) standardized against volumetric standard potassium iodate (article number 1.02404). The determined titer at 20°C was 1.000 with an expanded measurement uncertainty of ± 0.004 (k=2 coverage factor for 95% coverage probability). The certified value is traceable to primary standard NIST SRM 136f (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard potassium iodate, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.

Other Notes

Replaces: PX1443-6

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Potassium dichromate solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Liq. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Cardio-vascular system, Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library