Skip to Content
Merck
All Photos(1)

Key Documents

1.06601

Supelco

Sodium salicylate

for analysis EMSURE®

Synonym(s):

Sodium salicylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NaO3
CAS Number:
Molecular Weight:
160.10
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-198-0
NACRES:
NA.21
form:
solid
solubility:
1000 g/L
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 25 °C)

Quality Level

product line

EMSURE®

Assay

≥99.5% dry basis (perchloric acid titration)

form

solid

autoignition temp.

>250 °C

potency

930 mg/kg LD50, oral (Rat)

impurities

≤0.005% In water insoluble matter
≤0.2% Water

pH

6.5 (20 °C, 100 g/L in H2O)

mp

213 °C

solubility

1000 g/L

density

0.32 g/cm3 at 20 °C

bulk density

320 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

storage temp.

15-25°C

SMILES string

[Na+].[O-]C(=O)c1c(cccc1)O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Improving Detection of Amplitude-Modulated Sound: Sodium salicylate has been shown to enhance the detection of amplitude-modulated sound in mice, suggesting its potential use in auditory research and therapies aimed at improving sound perception and processing (van den Berg et al., 2024).

  • Grain Protein Content Determination: In agricultural chemistry, sodium salicylate is employed in the indophenol blue colorimetric method to determine the grain protein content of cereal plants, showcasing its application in enhancing agricultural productivity and food quality assessment (Huang et al., 2024).

Linkage

Replaces: SX0745; SX0745-1; SX0745-3

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.5 %
Identity: passes test
Appearance of solution (10 %; water): passes test
In water insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.002 %
Sulphate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Water: ≤ 0.2 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service