Skip to Content
Merck
All Photos(1)

Key Documents

1.06557

Supelco

di-Sodium oxalate

for analysis EMSURE®

Synonym(s):

di-Sodium oxalate, Oxalic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2Na2O4
CAS Number:
Molecular Weight:
134.00
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-550-3
NACRES:
NA.21
Assay:
≥99.8% (manganometric)
form:
solid
solubility:
37 g/L

Quality Level

product line

EMSURE®

Assay

≥99.8% (manganometric)

form

solid

potency

375 mg/kg LD50, oral (Rat)

loss

≤0.05% loss on drying, 105°C

pH

8 (20 °C, 30 g/L in H2O)

mp

250-270 °C (decomposition)

solubility

37 g/L

density

2.27 g/cm3 at 20 °C

bulk density

600 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤0.0005%
K: ≤0.005%
N: ≤0.001%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]C(=O)C(=O)[O-]

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (manganometric): ≥ 99.8 %
pH-value (3 %, water): 7.5 - 8.5
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.002 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Loss on Drying (105°C): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service