Skip to Content
Merck
All Photos(1)

Key Documents

1.04905

Supelco

Potassium bromide

for analysis (max. 0,000001% Hg) EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Potassium bromide, Hydrobromic Acid Potassium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BrK
CAS Number:
Molecular Weight:
119.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-830-3
NACRES:
NA.21
Assay:
≥99.5% (silver nitrate titration)
99.5-100.5% dry basis (silver nitrate titration)
grade:
ACS reagent
form:
solid
solubility:
650 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

1.3 hPa ( 795 °C)

product line

EMSURE®

Assay

≥99.5% (silver nitrate titration)
99.5-100.5% dry basis (silver nitrate titration)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

loss

≤0.5% loss on drying, 105°C

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

bp

1435 °C/1013 hPa

mp

730 °C

solubility

650 g/L

density

2.75 g/cm3 at 20 °C

bulk density

900‑1000 kg/m3

anion traces

bromate (BrO3-): ≤0.001%
bromate (BrO3-): passes test
chloride (Cl-): ≤0.1%
iodate (IO3-): ≤0.001%
iodide (I-): ≤0.001%passes test
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.001%
Fe: ≤0.0005%
Hg: ≤0.000001%
Mg: ≤0.001%
Mg: ≤0.02% (alkaline-earth metals (as Ca))
Na: ≤0.02%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Transition of Psychotropic Drugs in Japanese Pharmacopoeia.: Discusses the evolution of the standards and test methods for Potassium Bromide, comparing Japanese Pharmacopoeia to USP and BP (Yanagisawa K, 2015).
  • False hyperchloraemia in a dog due to potassium bromide.: Investigates an analytical chemistry error caused by potassium bromide in a veterinary context (Peacock RE et al., 2013).
  • Standard and real patient kidney stone library using FTIR spectroscopy.: Utilizes potassium bromide in the development of a standard library for kidney stones using Fourier transform-infrared spectroscopy (Mulready KJ et al., 2012).
  • Quantitative determination of solids by infrared spectroscopy.: Explores the use of potassium bromide disk technique for quantitative analysis in infrared spectroscopy (Jensen JB, 1958).
  • Research on quantitative determination of solids.: Continues the discussion on the quantitative determination using the potassium bromide disk technique in infrared spectroscopy (Jensen JB, 1958).

Linkage

Replaces: PX1380; PX1380-1

Analysis Note

Assay (argentometric): ≥ 99.5 %
Assay (argentometric; calculated on dried substance): 99.5 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
Acidity or alkalinity: passes test
pH-value (5 %; water): 5.0 - 8.0
Bromate(BrO₃): passes test
Bromate (BrO₃): ≤ 0.001 %
Chloride (Cl): ≤ 0.1 %
Iodate: ≤ 0.001 %
Iodide (I):
passes test
: ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ba (Barium): ≤ 0.002 %
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Hg (Mercury): ≤ 0.000001 %
Mg (Magnesium): ≤ 0.001 %
Na (Sodium): ≤ 0.02 %
Magnesium and alkaline-earth metals (as Ca): ≤ 0.02 %
Loss on Drying (105°C): ≤ 0.5 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service