Skip to Content

1.04401

Mercury

EMPLURA®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
12141726
eCl@ss:
38150701
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.0017 hPa ( 20 °C)

Quality Segment

assay

99.6-100.0% (complexometric)

form

liquid

impurities

≤0.002% Substances insoluble in nitric acid

bp

357 °C/1013 hPa

mp

-39 °C

solubility

0.06 mg/L

density

13.55 g/cm3 at 20 °C

storage temp.

20-25°C

Analysis Note

Assay (complexometric):99.6 - 100.0 %
Appearance of solution:passes test
Identity:passes test
Substances insoluble in nitric acid:≤ 0.002 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Still not finding the right product?

Explore all of our products under Mercury


Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library