Skip to Content
Merck

1.03252

2,2-Bis(hydroxyethyl)-(iminotris)- (hydroxymethyl)-methane

BIS-TRIS

Synonym(s):

2,2-Bis(hydroxyethyl)-(iminotris)- (hydroxymethyl)-methane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
MDL number:
UNSPSC Code:
12352104
EC Index Number:
230-237-7
NACRES:
NA.05
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

pH

9.4-9.8 (20 °C, 20 g/L in H2O)

solubility

soluble 579.9 g/L

storage temp.

15-25°C

SMILES string

N(C(CO)(CO)CO)(CCO)CCO

InChI

1S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14H,1-7H2

InChI key

OWMVSZAMULFTJU-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 99.0 %
Identity (IR-spectrum): conforms
Absorbance A (280 nm; 10 %; 1 cm; water): ≤ 0.1
pH-value (2 %; water): 9.4 - 9.8
Chloride (Cl): ≤ 0.005 %
Sulphate (SO₄): ≤ 0.01 %
Fe (Iron): ≤ 0.001 %
Pb (Lead): ≤ 0.001 %
Sulfated ash: ≤ 0.05 %
Loss on drying (60 °C; vacuum): ≤ 1.0 %


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library