Skip to Content
Merck

1.02452

L-Cysteine

for cell culture media

Synonym(s):

L-Cysteine, α-Amino-β-mercapto propionic acid, αlpha-Amino-β-mercapto propionic acid, Cys, αlpha-Amino-beta-mercapto propionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C3H7NO2S
CAS Number:
Molecular Weight:
121.16
UNSPSC Code:
41106213
EC Index Number:
200-158-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

solid

autoignition temp.

420 °C

potency

1890 mg/kg LD50, oral (Rat)

pH

4.5-5.5 (20 °C, 100 g/L in H2O)

mp

220-228 °C

solubility

280 g/L

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

SCC([N+H3])C(=O)[O-]

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)

InChI key

XUJNEKJLAYXESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
102415000004054839634727
102452025004027269965280
102452002504027269669775

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service