Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
UNSPSC Code:
12141707
EC Index Number:
231-152-8
NACRES:
NA.21
MDL number:
vapor pressure
1.3 hPa ( 394 °C)
Quality Level
product line
EMSURE®
form
solid
impurities
≤0.0005% Hg (Mercury), ≤0.001% Cu (Copper), ≤0.001% Fe (Iron), ≤0.001% Zn (Zinc), ≤0.005% Pb (Lead)
mp
321 °C
solubility
insoluble 8.2 mg/L
density
8.6 g/cm3 at 22 °C
storage temp.
no temp limit
SMILES string
[Cd+2]
InChI
1S/Cd/q+2
InChI key
WLZRMCYVCSSEQC-UHFFFAOYSA-N
Analysis Note
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Hg (Mercury): ≤ 0.0005 %
Pb (Lead): ≤ 0.005 %
Zn (Zinc): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Hg (Mercury): ≤ 0.0005 %
Pb (Lead): ≤ 0.005 %
Zn (Zinc): ≤ 0.001 %
Legal Information
EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Cadmium
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1020040250 | 04022536027487 |
| 1020040000 | 04022536027470 |
