Skip to Content
Merck

1.02001

Cadmium

coarse powder, for analysis and for filling reductors particle size about 0.3-1.6 mm EMSURE®

Synonym(s):

Cadmium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
UNSPSC Code:
12141707
EC Index Number:
231-152-8
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1.3 hPa ( 394 °C)

Quality Level

product line

EMSURE®

form

solid

impurities

≤0.001% Fe (Iron), ≤0.002% Cu (Copper), ≤0.005% Zn (Zinc), ≤0.01% Pb (Lead)

mp

321 °C

density

8.6 g/cm3 at 22 °C

storage temp.

2-30°C

SMILES string

[Cd+2]

InChI

1S/Cd/q+2

InChI key

WLZRMCYVCSSEQC-UHFFFAOYSA-N

Analysis Note

Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
Pb (Lead): ≤ 0.01 %
Zn (Zinc): ≤ 0.005 %

Other Notes

Replaces: 2001-4; 2001; 2001-2

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Cadmium


signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
102001025004022536027371
102001100704027269164799
102001100004022536027388
102001000004022536027364