Skip to Content
Merck

1.01960

Potassium dihydrogen phosphate/ di-sodium hydrogen phosphate

certified reference material, traceable to NIST (SRM), traceable to PTB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

traceable to NIST (SRM), traceable to PTB

storage temp.

15-25°C

General description

Certipur® certified secondary standard reference materials are used to precisely calibrate and monitor pH measurement instruments and buffer solutions. They are analyzed in our ISO/IEC 17025 and ISO 17034 accredited calibration laboratories against certified primary standard reference materials (SRM) in accordance with DIN 19266, IUPAC, NIST, Ph. Eur and USP. Certipur® reference materials offer maximum accuracy, reliability and convenience and help avoid costly repeat analyses.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Potassium dihydrogen phosphate/ di-sodium hydrogen phosphate [pH(S)=6.86₅ /pH(S)=7.41₃ (25 °C)] is a certified secondary standard buffer solution used as a calibration buffer standard for pH instruments or pH electrodes.

Features and Benefits

  • The product is delivered with a comprehensive, batch-specific Certificate of Analysis showing the relevant data for the quality management system.
  • The pH value of this Certipur® buffer solution is qualified using differential potentiometry with two platinum hydrogen electrodes against NIST & PTB primary reference materials [pH(S)=6.865 /pH(S)=7.413 (25 °C))].
  • This buffer standard has a guaranteed shelf life of 4 years.
  • It is a Certified Reference Material (CRM) offered in neat format. Solution of this standard is prepared in-house per instructions in European Pharmacopoeia (Ph. Eur. 2.2.3).
  • Intended for extremely precise calibration, qualification and monitoring of pH instruments and routine assessment of their performance.

Analysis Note

pH-value (25 °C): 6.861 - 6.867

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
101960000104022536026923