Skip to Content
Merck

1.01746

Sulfite Test, photometric

photometric, 1.0-60.0 mg/L (SO32-), 0.8-48.0 mg/L (SO2), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
eCl@ss:
40050204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sulfite Test, photometric, 1.0-60.0 mg/L (SO32-), 0.8-48.0 mg/L (SO2), Spectroquant®

product line

Spectroquant®

Quality Segment

usage

sufficient for 150 tests

specific analyte(s)

sulfite

measuring range

0.8-48.0 mg/L (SO2), 1.0-60.0 mg/L (SO32-)

technique(s)

photometry: suitable

application(s)

beverage testing
boiler and cooling water
cannabis testing
food testing
ground and surface water
mineral water
wastewater

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

detection method

photometric (Ellman’s reagent)

storage temp.

15-25°C

General description

Method: photometric 1.0 - 60.0 mg/l SO₃²⁻ 0.8 - 48.0 mg/l SO₂ Spectroquant®

This Spectroquant® Sulfite Reagent Test allows the accurate quantification of the sulfite content in aqueous samples as well as in food and beverages (e.g. beer).

Method applied: In neutral solution, sulfite ions react with 2,2’-dinitro-5,5’-dithiodibenzoic acid (Ellman’s reagent) to form an organic thiosulfate. This reaction results in the release of a thiol which is determined photometrically.

The Spectroquant®Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class

10 - Combustible liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library