Skip to Content
Merck

Skip To

1.00820

Methenamine silver plating kit acc. to Gomori

for the detection of argent-affine structures in histological tissue

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
41116124
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology

storage temp.

15-25°C

General description

The Methenamine silver plating kit acc. to Gomori - for the detection of argent-affine structures in histological tissue, is used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin, for example histological sections of e. g. the kidney, the intestine, or the liver. The periodic acid Schiff (PAS) reaction is often used for the visualization of basal membranes, fungal elements, or other argentaffin structures in the tissue. However, the Methenamine silver plating acc. to Gomori results in better and richer-in-contrast visualization of these target structures.
In the first step of the silver plating, the Periodic acid oxidizes the 1,2-glycols in the tissue to aldehydes. Due to the incubation with Methenamin-borat silver nitrate solution, these aldehydes are reduced and in turn are responsible for the reduction of the silver ions to metallic silver. This silver will appear black. The subsequent treatment of the tissue with gold chloride intensifies the staining of the argent-affinic structures and concurrently reduces the background staining, and the washing with sodium thiosulfate solution will remove the excess of silver-gold complexes. Like this, the target structures will exhibit a dark brown to black color and all other structures will be counter-stained with light green SF.
The package is sufficient for up to 50 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.002511.003621.00485
application(s)

clinical testing
diagnostic assay manufacturing
hematology

application(s)

clinical testing
diagnostic assay manufacturing
hematology, histology

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

15-25°C

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Thyroid

wgk

WGK 3

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters