Select a Size
About This Item
Product Name
Sulfuric acid 95-97%, for analysis (max. 0.005 ppm Hg) EMSURE® ACS,ISO,Reag. Ph Eur
grade
ACS reagent
Quality Level
agency
reag. ISO, reag. Ph. Eur.
vapor density
<0.3 (25 °C, vs air)
vapor pressure
0.0001 hPa ( 20 °C), 1 mmHg ( 146 °C)
product line
EMSURE®
assay
95-97% (alkalimetric)
form
liquid
ign. residue
≤5 ppm
color
APHA: ≤10
pH
0.3 (25 °C, 49 g/L in H2O)
bp
~290 °C (lit.)
mp
-20 °C
solubility
water: soluble (caution ! development of heat)
density
1.840 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤0.1 ppm, nitrate (NO3-): ≤0.2 ppm, phosphate (PO43-): ≤0.5 ppm
SMILES string
OS(O)(=O)=O
cation traces
Ag: ≤0.02 ppm, Al: ≤0.05 ppm, As: ≤0.01 ppm, Ba: ≤0.05 ppm, Be: ≤0.01 ppm, Bi: ≤0.1 ppm, Ca: ≤0.2 ppm, Cd: ≤0.02 ppm, Co: ≤0.01 ppm, Cr: ≤0.05 ppm, Cu: ≤0.01 ppm, Fe: ≤0.1 ppm, Ge: ≤0.05 ppm, Hg: ≤0.005 ppm, K: ≤0.1 ppm, Li: ≤0.01 ppm, Mg: ≤0.05 ppm, Mn: ≤0.01 ppm, Mo: ≤0.05 ppm, NH4+: ≤2 ppm, Na: ≤0.5 ppm, Ni: ≤0.02 ppm, Pb: ≤0.02 ppm, Sr: ≤0.02 ppm, Ti: ≤0.1 ppm, Tl: ≤0.05 ppm, V: ≤0.01 ppm, Zn: ≤0.05 ppm, Zr: ≤0.1 ppm, heavy metals (as Pb): ≤1 ppm
storage temp.
no temp limit
InChI
1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)
InChI key
QAOWNCQODCNURD-UHFFFAOYSA-N
Application
- Nuclear magnetic resonance analysis of a chemically cross-linked ferrous-methylthymol blue-polyvinyl alcohol radiochromic gel dosimeter.: Researchers applied NMR to analyze a ferrous-methylthymol blue-polyvinyl alcohol gel dosimeter, showing its potential for accurate radiation dose measurements in medical and industrial applications (Eyadeh et al., 2019).
- Discriminate sensing of pyrophosphate using a new tripodal tetramine-based dinuclear Zn(II) complex under an indicator displacement assay approach.: This work presented a novel sensing method for pyrophosphate detection using a Zn(II) complex and methylthymol blue in an indicator displacement assay, highlighting its selectivity and efficiency (Watchasit et al., 2014).
- Direct colorimetric detection of copper(II) ions in sampling using diffusive gradients in thin-films.: The study introduced a colorimetric technique utilizing methylthymol blue for direct detection of copper(II) ions via diffusive gradients in thin-films, offering a simple and effective analytical tool (McGifford et al., 2010).
- Experimental correlation between the pKa value of sulfonphthaleins with the nature of the substituents groups.: This research explored the relationship between the pKa values of sulfonphthaleins, including methylthymol blue, and their substituent groups, providing insights for better understanding and predicting their behavior in various chemical environments (Balderas-Hernández et al., 2008).
Analysis Note
Identity: passes test
Color: ≤ 10 Hazen
Appearance: passes test
Density (d 20/20): 1.834 - 1.837
Chloride (Cl): ≤ 0.1 ppm
Nitrate (NO₃): ≤ 0.2 ppm
Phosphate (PO₄): ≤ 0.5 ppm
Heavy metals (as Pb): ≤ 1 ppm
Ag (Silver): ≤ 0.02 ppm
Al (Aluminium): ≤ 0.05 ppm
As (Arsenic): ≤ 0.01 ppm
Ba (Barium): ≤ 0.05 ppm
Be (Beryllium): ≤ 0.01 ppm
Bi (Bismuth): ≤ 0.1
Ca (Calcium): ≤ 0.2 ppm
Cd (Cadmium): ≤ 0.02 ppm
Co (Cobalt): ≤ 0.01 ppm
Cr (Chromium): ≤ 0.05 ppm
Cu (Copper): ≤ 0.01 ppm
Fe (Iron): ≤ 0.1
Ge (Germanium): ≤ 0.05 ppm
Hg (Mercury): ≤ 0.005 ppm
K (Potassium): ≤ 0.1
Li (Lithium): ≤ 0.01 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.01 ppm
Mo (Molybdenum): ≤ 0.05 ppm
NH₄ (Ammonium): ≤ 2 ppm
Na (Sodium): ≤ 0.5 ppm
Ni (Nickel): ≤ 0.02 ppm
Pb (Lead): ≤ 0.02 ppm
Sr (Strontium): ≤ 0.02 ppm
Ti (Titanium): ≤ 0.1
Tl (Thallium): ≤ 0.05 ppm
V (Vanadium): ≤ 0.01 ppm
Zn (Zinc): ≤ 0.05 ppm
Zr (Zirconium): ≤ 0.1
Substances reducing potassium permanganate (as SO₂): ≤ 2 ppm
Residue on ignition: ≤ 5 ppm
Date of expiry: see product label
Legal Information
Still not finding the right product?
Explore all of our products under Sulfuric acid 95-97%
signalword
Danger
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
hcodes
Storage Class
8B - Non-combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1007321000 | 04027269568733 |
| 1007320510 | 04054839164187 |
| 1007322500 | 04022536009971 |
| 1007322510 | 04027269568986 |
| 1007329025 | 04022536779638 |
