Skip to Content

1.00677

AOX Sample Preparation Set

sufficient for 25 tests

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 25 tests

Quality Segment

analyte functional class(es)

adsorbable organic halides (AOX)

technique(s)

photometry: suitable

application(s)

boiler and cooling water
disposal drainage water
drinking water
electroplating
environmental
ground and surface water
mineral water
oil and petrochemicals
wastewater

storage temp.

15-25°C

General description

This AOX Sample Preparation Set is a required accessory when performing the Spectroquant® AOX Cell Test Kit 1.00675. It contains all necessary reagents and equipment for sample preparation:
1 bottle each of reagents AOX-1, AOX-2, AOX-3, and AOX-4
2 bottles of reagent AOX-5
1 bottle of reagent AOX-6
1 blue microspoon
25 adsorption columns with stoppers
1 graduated 12-ml plastic syringe with connector
More information can be found in the instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Ox. Liq. 3 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library