Skip to Content
Merck

1.00663

Parteck® LUB MST

(MAGNESIUM STEARATE VEGETABLE GRADE), EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF, FCC

Synonym(s):

PARTECK LUB MST

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H70MgO4
CAS Number:
Molecular Weight:
591.24
UNSPSC Code:
12352107
EC Index Number:
209-150-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, NF, Ph. Eur.

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

fine powder

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Olefin Metathesis

particle size

3-7 μm (d50)

pH

7 ( in H2O, slurry)

mp

140 °C

application(s)

pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[Mg+2].[O-]C(=O)CCCCCCCCCCCCCCCCC.[O-]C(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Mg/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

HQKMJHAJHXVSDF-UHFFFAOYSA-L

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® LUB, a product line of our most effective excipients for lubrication, demonstrates excellent batch-to-batch consistency and helps you to achieve a reliable workflow in your tablet manufacture. Parteck® LUB MST is a vegetable-origin magnesium stearate.

Features and Benefits

  • Consistent functionality and performance
  • Vegetable origin for minimal regulatory effort
  • Specified particle size and surface area for a reliable performance

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.