Skip to Content

1.00616

Phosphate Cell Test, photometric

photometric, 3.0-100.0 mg/L (PO4-P), 9-307 mg/L (PO43-), 7-229 mg/L (P2O5), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate Cell Test, photometric, 3.0-100.0 mg/L (PO4-P), 9-307 mg/L (PO43-), 7-229 mg/L (P2O5), Spectroquant®

product line

Spectroquant®

Quality Segment

usage

sufficient for 25 tests

specific analyte(s)

phosphate

measuring range

3.0-100.0 mg/L (PO4-P), 7-229 mg/L (P2O5), 9-307 mg/L (PO43-)

technique(s)

photometry: suitable

application(s)

agriculture
biotechnology/fermenter
disposal drainage water
environmental
ground and surface water
seawater
wastewater

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

detection method

photometric (Phosphomolybdenum blue)

storage temp.

15-25°C

General description

(o-phosphate) Method: photometric, PMB 3.0 - 100.0 mg/l PO₄-P 9 - 307 mg/l PO₄³⁻ 7 - 229 mg/l P₂O₅ Spectroquant®
This Spectroquant® Phosphate Cell Test allows the accurate quantification of the orthophosphate content in aqueous samples.
In sulfuric solution orthophosphate ions react with molybdate ions to form molybdophosphoric acid. Ascorbic acid reduces this to phosphomolybdenum blue (PMB) that is determined photometrically.
The method is analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Phosphate Cell Test


pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

hcodes

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library