Skip to Content
Merck

1.00613

Nitrogen (total) Cell Test, photometric

photometric, 0.5-15.0 mg/L (N), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nitrogen (total) Cell Test, photometric, 0.5-15.0 mg/L (N), Spectroquant®

Quality Level

product line

Spectroquant®

usage

sufficient for 25 tests

specific analyte(s)

nitrogen

measuring range

0.5-15.0 mg/L (N)

technique(s)

photometry: suitable

application(s)

agriculture
aquaculture
boiler and cooling water
disposal drainage water
environmental
ground and surface water
milk and dairy products
wastewater

compatibility

for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

detection method

photometric (Koroleff digestion, 2,6-dimethylphenol)

storage temp.

15-25°C

General description

Method: photometric, DMP 0.5 - 15.0 mg/l N Spectroquant®

The method corresponds to ISO 23697-1, a norm standardizing the use of this "small-scale sealed tube method" for this important parameter. The digestion is analogous to EN ISO 11905-1. The determination of nitrate is analogous to DIN 38405-9.

This Spectroquant® Nitrogen (total) Cell Test allows the reliable determination of total nitrogen in water samples. Total nitrogen is the sum of total Kjeldahl nitrogen (organic nitrogen and ammonia), nitrate, and nitrite. Prior to the determination the sample needs to be digested in order to transfer organic and inorganic nitrogen containing compounds into nitrate that further reacts to a photometrically analyzable compound.

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Nitrogen (total) Cell Test


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

84.2 °F

flash_point_c

29 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100613000104022536733821