Skip to Content
Merck
All Photos(1)

Key Documents

1.00306

Supelco

Hydrobromic acid

47%, Suprapur®

Synonym(s):

Hydrogen bromide solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352301
NACRES:
NB.21

Product Name

Hydrobromic acid 47%, Suprapur®, for inorganic trace analysis

grade

for inorganic trace analysis

Quality Level

vapor pressure

10.6 hPa ( 20 °C)

Assay

47%

form

liquid

concentration

≥40-50% HBr

pH

(at 20 °C (68 °F)strongly acid)

bp

126 °C/1013 hPa

mp

-11 °C

solubility

soluble (Water: Soluble)

density

1.49 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤50000 ppb
iodide (I-): ≤ 10000 ppb
phosphate, phosphite (as PO43-): ≤50 ppb ppb
sulfate, sulfite (as SO42-): ≤500 ppb

cation traces

Ag: ≤1 ppb
Al: ≤10 ppb
As: ≤0.5 ppb
Au: ≤0.5 ppb
Ba: ≤0.5 ppb
Be: ≤0.5 ppb
Bi: ≤0.5 ppb
Ca: ≤10 ppb
Cd: ≤0.5 ppb
Co: ≤0.5 ppb
Cr: ≤0.5 ppb
Cu: ≤0.5 ppb
Fe: ≤5 ppb
Ga: ≤0.5 ppb
Ge: ≤0.5 ppb
Hg: ≤5 ppb
In: ≤0.5 ppb
K: ≤5 ppb
Li: ≤0.5 ppb
Mg: ≤1 ppb
Mn: ≤0.5 ppb
Mo: ≤0.5 ppb
Na: ≤20 ppb
Ni: ≤0.5 ppb
Pb: ≤0.5 ppb
Pd: ≤1 ppb
Sb: ≤0.5 ppb
Sn: ≤0.5 ppb
Sr: ≤0.5 ppb
Ti: ≤0.5 ppb
Tl: ≤0.5 ppb
U: ≤0.5 ppb
V: ≤0.5 ppb
Zn: ≤5 ppb
Zr: ≤0.5 ppb

SMILES string

Br

storage temp.

2-30°C

InChI

InChI=1S/BrH/H1h

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 47 %
Chloride (Cl): ≤ 50000 ppb
Iodide (I): ≤ 10000 ppb
Phosphate, Phosphite (as PO₄): ≤ 50 ppb
Sulfate and sulphite (as SO₄): ≤ 500 ppb
Ag (Silver): ≤ 1 ppb
Al (Aluminium): ≤ 10 ppb
As (Arsenic): ≤ 0.5 ppb
Au (Gold): ≤ 0.5 ppb
Ba (Barium): ≤ 0.5 ppb
Be (Beryllium): ≤ 0.5 ppb
Bi (Bismuth): ≤ 0.5 ppb
Ca (Calcium): ≤ 10 ppb
Cd (Cadmium): ≤ 0.5 ppb
Co (Cobalt): ≤ 0.5 ppb
Cr (Chromium): ≤ 0.5 ppb
Cu (Copper): ≤ 0.5 ppb
Fe (Iron): ≤ 5 ppb
Ga (Gallium): ≤ 0.5 ppb
Ge (Germanium): ≤ 0.5 ppb
Hg (Mercury): ≤ 5 ppb
In (Indium): ≤ 0.5 ppb
K (Potassium): ≤ 5 ppb
Li (Lithium): ≤ 0.5 ppb
Mg (Magnesium): ≤ 1 ppb
Mn (Manganese): ≤ 0.5 ppb
Mo (Molybdenum): ≤ 0.5 ppb
Na (Sodium): ≤ 20 ppb
Ni (Nickel): ≤ 0.5 ppb
Pb (Lead): ≤ 0.5 ppb
Pd (Palladium): ≤ 1 ppb
Sb (Antimony): ≤ 0.5 ppb
Sn (Tin): ≤ 0.5 ppb
Sr (Strontium): ≤ 0.5 ppb
Ti (Titanium): ≤ 0.5 ppb
Tl (Thallium): ≤ 0.5 ppb
U (Uranium): ≤ 0.5 ppb
V (Vanadium): ≤ 0.5 ppb
Zn (Zinc): ≤ 5 ppb
Zr (Zirconium): ≤ 0.5 ppb
Residue on ignition (as sulphate): ≤ 2000 ppb

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service