Skip to Content
Merck
All Photos(3)

Key Documents

09-843

Sigma-Aldrich

Anti-SIRT2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SIR2-like protein 2, silencing information regulator 2-like, silent information regulator 2, sir2-related protein type 2, sirtuin (silent mating type information regulation 2, homolog) 2 (S. cerevisiae)

sirtuin (silent mating type information r

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRT2(22933)

General description

Sirtuins are hypothesized to play a key role in an organism′s response to stresses (such as heat or starvation) and to be responsible for the lifespan-extending effects of calorie restriction. Sirtuins act by removing acetyl groups from proteins in the presence of NAD+; they are thus classified as "NAD+-dependent deacetylases". Mammals have seven sirtuins; SIRT1 and SIRT2 occupy the cytoplasm. SIRT2 is a NAD-dependent deacytylase. It has been shown to control gene silencing, cell cycle, and DNA damage repair. In additon, SIRT2 acts as a tumor suppressor in human gliomas which suggests that the ectopic expression of SIRT2 upsets the microtubule network.

Specificity

This antibody recognizes the mature protein of SIRT2.

Immunogen

Epitope: Mature protein
KLH-conjugated linear peptide corresponding to the mature protein of human SIRT2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Use Anti-SIRT2 Antibody (Rabbit Polyclonal Antibody) validated in WB, IP to detect SIRT2 also known as SIR2-like protein 2, silent information regulator 2, sirtuin 2.
Western Blot (SNAP ID) Analysis: 0.25 µg/mL from a representative lot detected SIRT2 on 10 µg of human brain tissue lysate.

Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated SIRT2 in human brain lysate.

Quality

Evaluated by Western Blot in human brain tissue lysate.

Western Blot Analysis: 0.125 µg/mL of this antibody detected SIRT2 on 10 µg of human brain tissue lysate.

Target description

Observed molecular weight is ~39 kDa Multiple isoforms around 40 kDa may be observed.

Linkage

Replaces: 04-1124

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Ellis et al.
Nature protocols, 16(2), 841-871 (2020-12-16)
Somatic mutations accumulate in healthy tissues as we age, giving rise to cancer and potentially contributing to ageing. To study somatic mutations in non-neoplastic tissues, we developed a series of protocols to sequence the genomes of small populations of cells
Roberta Scrimieri et al.
Biomedicines, 9(12) (2021-12-25)
Vitamin D (VitD) exerts protective effects on the endothelium, which is fundamental for vascular integrity, partly by inhibiting free radical formation. We found that VitD prevents high glucose-induced Thioredoxin Interacting Protein (TXNIP) upregulation. Increased amounts of TXNIP are responsible for
Klara Metzner et al.
Frontiers in cell and developmental biology, 10, 965382-965382 (2022-11-18)
Aging is a complex process characterized by several molecular and cellular imbalances. The composition and stability of the neuronal cytoskeleton is essential for the maintenance of homeostasis, especially in long neurites. Using human skin biopsies containing sensory axons from a
Jonathan P Belman et al.
The Journal of biological chemistry, 290(7), 4447-4463 (2015-01-07)
Insulin causes the exocytic translocation of GLUT4 glucose transporters to stimulate glucose uptake in fat and muscle. Previous results support a model in which TUG traps GLUT4 in intracellular, insulin-responsive vesicles termed GLUT4 storage vesicles (GSVs). Insulin triggers TUG cleavage
Sung Eun Wang et al.
Experimental neurobiology, 28(4), 537-546 (2019-09-09)
Silent information regulator 2 (Sirtuin2 / SIRT2) is a NAD+-dependent deacetylase that regulates the cellular oxidative stress response. It modulates transcriptional silencing and protein stability through deacetylation of target proteins including histones. Previous studies have shown that SIRT2 plays a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service