Skip to Content
Merck
All Photos(6)

Key Documents

09-482

Sigma-Aldrich

Anti-PI3 Kinase Antibody, p110β

from rabbit, purified by affinity chromatography

Synonym(s):

PI3-kinase p110 subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, PtdIns-3-kinase p110, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, ph

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, canine

species reactivity (predicted by homology)

rat (immunogen homology), mouse (immunogen homology), dog (immunogen homology), monkey (immunogen homology)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PIK3CB(5291)

General description

PI3 Kinase (Phosphatidylinositol 3-Kinase) is responsible for phosphorylation of the 3 position of the inositol ring of PI(4,5)P2, to generate PI(3,4,5)P3, a potent second messenger required for survival, signaling, and insulin action. PI3 Kinase is a heterodimeric complex composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. Tyrosine phosphorylation of growth factor receptors creates docking sites for binding of p85 (through its SH2 domains) on the receptors; p85 brings with it p110, which is then proximal to its phospho-lipid substrate on the membrane. PI3 Kinase is also activated by Ras, and by the β:γ subunits of heterotrimeric G-proteins. PI3 Kinase is inhibitable by wortmannin, a useful tool for the study of the PI3 Kinase signaling pathway.

Specificity

Recognizes the internal domain of PI3 Kinase, p110β.

Immunogen

Epitope: Internal Domain
KLH-conjugated synthetic linear peptide corresponding to the internal domain of PI3 Kinase, p110β.

Application

Anti-PI3 Kinase Antibody, p110β is an antibody against PI3 Kinase for use in WB, IP, IC, IH, ELISA.
ELISA:
This antibody was tested and determined to work for ELISA at 0.5 ng/well.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Evaluated by Western Blot on HEK-293 cell lysate.
Western Blot Analysis: 0.05 - 0.1 μg/mL of this antibody detected endogenous PI3 Kinase, p110β in lysate from HEK-293 cells.

Target description

110 kDa

Linkage

Replaces: 04-399

Physical form

Affinity purified
Antigen affinity purified rabbit serum in 0.1M Tris-Glycine (pH7.4), 150mM NaCl, with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Analysis Note

Control
HEK-293 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mobilization of mesenchymal stem cells by stromal cell-derived factor-1 released from chitosan/tripolyphosphate/fucoidan nanoparticles.
Yi-Cheng Huang,Ting-Ju Liu
Acta Biomaterialia null
Pattaraporn Chaisupasakul et al.
PloS one, 19(3), e0300051-e0300051 (2024-03-25)
The cytotoxicity of the ethyl acetate fraction of the Calotropis gigantea (L.) Dryand. (C. gigantea) stem bark extract (CGEtOAc) has been demonstrated in many types of cancers. This study examined the improved cancer therapeutic activity of sorafenib when combined with
Ashwini C Poopal et al.
Molecular autism, 7, 3-3 (2016-01-16)
Dysfunctions in the PI3K/mTOR pathway have gained a lot of attention in autism research. This was initially based on the discovery of several monogenic autism spectrum disorders with mutations or defects in PI3K/mTOR signaling components. Recent genetic studies corroborate that
Orkide Palabiyik et al.
Biomedical reports, 0(0), 1-10 (2019-03-08)
Athletes misuse recombinant human growth hormone (r-hGH) to enhance their performance. Although r-hGH is known to increase cardiac hypertrophy, the underlying molecular mechanism remains unclear. The aim of the present study was to investigate the role of r-hGH in cardiac
Sabiha Abekhoukh et al.
Disease models & mechanisms, 10(4), 463-474 (2017-02-12)
Cytoplasmic FMRP interacting protein 1 (CYFIP1) is a candidate gene for intellectual disability (ID), autism, schizophrenia and epilepsy. It is a member of a family of proteins that is highly conserved during evolution, sharing high homology with its Drosophila homolog

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service