Skip to Content
Merck

09-217

Anti-Raptor Antibody

Upstate®, from rabbit

Synonym(s):

p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, regulatory associated protein of mTOR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RPTOR(57521)

General description

150 kDa

Immunogen

KLH conjugated peptide encompassing the N-terminal region of human Raptor.

Application

Detect Raptor using this anti-Raptor Antibody validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Biochem/physiol Actions

Other species have not been tested.

Physical form

Affinity purified rabbit polyclonal IgG in 0.02M PBS, 0.25M sodium chloride containing 0.1% sodium azide and 50% glycerol.
ImmunoAffinity Purified

Preparation Note

2 years at -20°C from date of shipment

Analysis Note

Routinely evaluated by immunoblot.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library