Skip to Content
Merck
All Photos(1)

Key Documents

07-710

Sigma-Aldrich

Anti-SMC2/hCAP-E Antibody

serum, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SMC2(10592)

General description

SMC2 and SMC4 from the active ATPase subunit of Condensin I and Condensin II complexes that regulate chromosome compaction.

Specificity

SMC2/hCAP-E

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 1184-1197 (C-KSKAKPPKGAHVEV) of human SMC2/hCAP-E (Structural Maintenance of Chromosome 2/ Chromosome-Associated Protein E) with an N-terminal cysteine added for conjugation purposes.

Application

Anti-SMC2/hCAP-E Antibody is a high quality Rabbit Polyclonal Antibody for the detection of SMC2/hCAP-E & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Routinely evaluated by immunoblot.

Target description

137kDa

Physical form

Serum

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Classification, subtype discovery, and prediction of outcome in pediatric acute lymphoblastic leukemia by gene expression profiling.
Yeoh, Eng-Juh, et al.
Cancer Cell, 1, 133-143 (2002)
Alexander Schleiffer et al.
Molecular cell, 11(3), 571-575 (2003-04-02)
We describe a superfamily of eukaryotic and prokaryotic proteins (kleisins) that includes ScpA, Scc1, Rec8, and Barren. Scc1 interacts with SMC proteins through N- and C-terminal domains to form a ring-like structure. Since these are the only domains conserved among
Mingxuan Sun et al.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology, 26(4), 277-295 (2018-08-26)
During cell division, chromosomes must be folded into their compact mitotic form to ensure their segregation. This process is thought to be largely controlled by the action of condensin SMC protein complexes on chromatin fibers. However, how condensins organize metaphase
K Kimura et al.
The Journal of biological chemistry, 276(8), 5417-5420 (2001-01-04)
13S condensin is a five-subunit protein complex that plays a central role in mitotic chromosome condensation. The condensin complex was originally identified and purified from Xenopus egg extracts and shown to have an ATP-dependent positive supercoiling activity in vitro. We
J A Schmiesing et al.
Molecular and cellular biology, 20(18), 6996-7006 (2000-08-25)
Structural maintenance of chromosomes (SMC) family proteins play critical roles in structural changes of chromosomes. Previously, we identified two human SMC family proteins, hCAP-C and hCAP-E, which form a heterodimeric complex (hCAP-C-hCAP-E) in the cell. Based on the sequence conservation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service