Recommended Products
biological source
rabbit
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... UTY(7404)
Specificity
Scrib
Immunogen
fusion protein corresponding to residues 1595-1630 of human Scrib
Application
Detect Scrib using this Anti-Scrib Antibody validated for use in WB.
Research Category
Signaling
Signaling
Research Sub Category
Growth Factors & Receptors
Growth Factors & Receptors
Quality
routinely evaluated by immunoblot from HeLa cells
Target description
220kDa
Physical form
70% storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide) and 30% glycerol.
Format: Purified
Protein A purified
Storage and Stability
2 years at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Oncogene, 22(58), 9225-9230 (2003-12-19)
Scribble (scrib), discs large (dlg) and lethal giant larvae (lgl) encode proteins that regulate cell polarity and have been identified as neoplastic tumour suppressor genes in Drosophila melanogaster. Here, we have used the Drosophila model system to provide the first
Molecular and cellular biology, 20(21), 8244-8253 (2000-10-12)
The high-risk human papillomavirus (HPV) E6 proteins stimulate the ubiquitination and degradation of p53, dependent on the E6AP ubiquitin-protein ligase. Other proteins have also been shown to be targeted for degradation by E6, including hDlg, the human homolog of the
Cell cycle (Georgetown, Tex.), 7(11), 1613-1622 (2008-05-13)
Epithelial-to-mesenchymal transition (EMT) underlies cell plasticity and embryonic development and is frequently observed in advanced tumorigenesis. We demonstrated that midkine (MK), a retinoic acid-inducible heparin-binding mitogen, promotes EMT in immortalized HaCaT keratinocytes. We showed that MK binds to the Notch2
Current opinion in cell biology, 16(5), 493-499 (2004-09-15)
The size of epithelial tubes is critical for the function of organs such as the lung, kidney and vascular system. However, the molecular mechanisms regulating tube size are largely unknown. Recent work in the Drosophila tracheal system reveals that septate
BioEssays : news and reviews in molecular, cellular and developmental biology, 25(6), 542-553 (2003-05-27)
Dlg (Discs large), Scrib (Scribble) and Lgl (Lethal giant larvae) are evolutionarily conserved components of a common genetic pathway that link the seemingly disparate functions of cell polarity and cell proliferation in epithelial cells. dlg, scrib and lgl have been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service