Skip to Content

07-633

Anti-HNF3β/FOXA2 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-HNF-3-beta, Anti-HNF3B, Anti-TCF3B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Species reactivity:
mouse, human, rat
Technique(s):
immunohistochemistry: suitable, western blot: suitable
Application:
IHC, WB
Citations:
77
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXA2(3170)
mouse ... Foxa2(15376)
rat ... Foxa2(25099)

General description

50 kDa
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. This gene has been linked to sporatic cases of maturity-onset diabetes of the young. Two transcript variants encoding the same protein have been identified for this gene.

Immunogen

GST fusion protein corresponding to residues 7-86 of rat hepatocyte nuclear factor-3 beta / forkhead box A2 (HNF3β/FOXA2)

Application

Detect HNF3β/FOXA2 with Anti-HNF3β/FOXA2 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in IHC & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Biochem/physiol Actions

Recognizes HNF3β/FOXA2.

Physical form

Format: Purified
Protein A Purified rabbit IgG in a solution of 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.135M NaCl, and 0.035% sodium azide as a preservative.
Protein A purified

Preparation Note

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
MLE 12 nuclear extracts, human liver lysate
Routinely evaluated by immunoblot with nuclear extracts from MLE 12 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library