Skip to Content
Merck

07-594

Anti-Histone H2A.Z Antibody

serum, Upstate®

Synonym(s):

Anti-H2A.Z-1, Anti-H2A.z, Anti-H2A/z, Anti-H2AFZ, Anti-H2AZ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
28
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... H2AFZ(3015)

General description

16kDa

Immunogen

peptide containing the sequence SLIGKKGQQ, corres-ponding to the C-terminus of human histone H2A.Z

Application

Detect Histone H2A.Z also known as H2AZ histone with Anti-Histone H2A.Z Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

Broad species cross-reactivity is expected
C-terminus of histone H2A.Z

Physical form

0.05% sodium azide before the addition of glycerol to 30%
Serum

Preparation Note

2 years at -20°C

Analysis Note

routinely evaluated by immunoblot on acid extracted proteins from HeLa cells and recombinant Histone H2A.Z (Catalog #14-597), no cross-reactivity with Histone H2A

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library