Skip to Content
Merck

Skip To

07-455

Anti-CoREST Antibody

Upstate®, from rabbit

Synonym(s):

Anti-COREST, Anti-RCOR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, ICC, IHC, IP, WB
Citations:
68
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RCOR1(23186)

General description

66 kDa
The CoREST gene encodes a functional corepressor required for regulation of neural specific gene expression. REST (RE1 silencing transcription factor 1) can mediate extraneuronal restriction by imposing either active repression via histone deacetylase recruitment or long term gene silencing using a distinct functional complex. Silencing of neuronal specific genes requires the recruitment of an associated corepressor, CoREST, that serves as a functional molecular beacon for the recruitment of molecular machinery that imposes silencing across a chromosomal interval, including transcriptional units that do not themselves contain REST response elements.

Immunogen

GST fusion protein corresponding to residues 109-293 of human CoREST.

Application

Detect CoREST with Anti-CoREST Antibody (Rabbit Polyclonal Antibody), that has been shown to work in ChIP, ICC, IHC, IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Biochem/physiol Actions

Recognizes CoREST.

Physical form

Format: Purified
Protein A Purified immunoglobulin in 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, and 0.05% sodium azide as a preservative.
Protein A purified

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.
routinely evaluated by immunoblot in RIPA lysates from A431, 3T3/A31, Jurkat,and Hela cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABN486MABE570ABE365
species reactivity

rat, human, mouse

species reactivity

rat, human, mouse

species reactivity

human

species reactivity

human, mouse, rat

clone

polyclonal

clone

K72/8, monoclonal

clone

7A7A9, monoclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

affinity isolated antibody

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

rabbit

technique(s)

ChIP: suitable, immunohistochemistry: suitable, western blot: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

immunohistochemistry: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters