Skip to Content
Merck

07-356

Anti-NR3A Antibody

Upstate®, from rabbit

Synonym(s):

Anti-GluN3A, Anti-NMDAR-L, Anti-NMDAR3A, Anti-NR3A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
38
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN3A(116443)

General description

130kDa

Immunogen

peptide corresponding to amino acids 1098-1113 (SRKTELEEYQKTNRTC) of rat NR3A

Application

Anti-NR3A Antibody is an antibody against NR3A for use in WB.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Neurodegenerative Diseases

Biochem/physiol Actions

NR3A

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Preparation Note

2 years at -20°C

Analysis Note

routinely evaluated by immunoblot in a rat brain microsomal protein preparation

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library