Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
150 μL | Check Cart for Availability | $476.00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Jak3(16453)
General description
Immunogen
Application
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone B32-32, monoclonal | clone polyclonal | clone polyclonal |
| antibody form serum | antibody form purified antibody | antibody form serum | antibody form serum |
| species reactivity mouse | species reactivity human, mouse | species reactivity rabbit, hamster, mouse, human, rat | species reactivity mouse, human, rat |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Explore all of our products under Anti-JAK3 Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



