Skip to Content
Merck
All Photos(2)

Key Documents

06-332

Sigma-Aldrich

Anti-GST Tag Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

clone

polyclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Immunogen

GST (Glutathione-S-transferase) derived from the pGEX expression vector and linked to KLH

Application

Anti-GST Tag Antibody detects level of GST Tag & has been published & validated for use in WB.

Quality

routinely evaluated by immunoblot on MAP Kinase 2/Erk2, active (Catalog # 14-173). 0.5-2mg/ml of a previous lot detected 500ng of GST (Catalog # 12-350)

Target description

28-30 kDa

Physical form

Format: Purified

Analysis Note

Control
Cells expressing the GST-tag

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Brenna et al.
eLife, 8 (2019-11-07)
Circadian oscillations emerge from transcriptional and post-translational feedback loops. An important step in generating rhythmicity is the translocation of clock components into the nucleus, which is regulated in many cases by kinases. In mammals, the kinase promoting the nuclear import
Regulation of nuclear import/export of carbohydrate response element-binding protein (ChREBP): interaction of an alpha-helix of ChREBP with the 14-3-3 proteins and regulation by phosphorylation.
Sakiyama, H; Wynn, RM; Lee, WR; Fukasawa, M; Mizuguchi, H; Gardner, KH; Repa, JJ; Uyeda, K
The Journal of Biological Chemistry null
The telomeric protein TRF2 binds the ATM kinase and can inhibit the ATM-dependent DNA damage response.
Karlseder, J; Hoke, K; Mirzoeva, OK; Bakkenist, C; Kastan, MB; Petrini, JH; de Lange, T
PLoS Biology null
Lotus japonicus E3 ligase SEVEN IN ABSENTIA4 destabilizes the symbiosis receptor-like kinase SYMRK and negatively regulates rhizobial infection.
Den Herder, G; Yoshida, S; Antolin-Llovera, M; Ried, MK; Parniske, M
Plant Cell null
Laura Megrelis et al.
Frontiers in immunology, 9, 2001-2001 (2018-09-27)
We previously identified Fam65b as an atypical inhibitor of the small G protein RhoA. Using a conditional model of a Fam65b-deficient mouse, we first show that Fam65b restricts spontaneous RhoA activation in resting T lymphocytes and regulates intranodal T cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service