Skip to Content
Merck
All Photos(1)

Key Documents

05-782

Sigma-Aldrich

Anti-GST Tag Antibody

Upstate®, from mouse

Synonym(s):

Anti-DFN7, Anti-FAEES3, Anti-GST3, Anti-GSTP, Anti-HEL-S-22, Anti-PI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

bacteria

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

GST fusion proteins
Not expected to cross-react with GST from other species.

Immunogen

Recombinant GST (glutathione S-transferase) from Shistosoma japonicum expressed in E. coli

Application

Detect GST Tag using this Anti-GST Tag Antibody validated for use in WB.

Quality

routinely evaluated by immunoblot on recombinant GST protein (Catalog #12-350) and GST-Abltide fusion protein (Catalog #12-525)

Target description

varies depending upon the GST fusion proteins being recognized

Physical form

Format: Purified
protein G purified mouse IgG in storage buffer (PBS containing 0.05% sodium azide) and 30% glycerol

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuning microtubule-based transport through filamentous MAPs: the problem of dynein.
Michael Vershinin, Jing Xu, David S Razafsky, Stephen J King, Steven P Gross
Traffic null
Malin H Stridh et al.
The Journal of physiology, 590(10), 2333-2351 (2012-03-28)
Rapid exchange of metabolites between different cell types is crucial for energy homeostasis of the brain. Besides glucose, lactate is a major metabolite in the brain and is primarily produced in astrocytes. In the present study, we report that carbonic
Stuart Sullivan et al.
Nature communications, 12(1), 6129-6129 (2021-10-23)
Polarity underlies all directional growth responses in plants including growth towards the light (phototropism). The plasma-membrane associated protein, NON-PHOTOTROPIC HYPOCOTYL 3 (NPH3) is a key determinant of phototropic growth which is regulated by phototropin (phot) AGC kinases. Here we demonstrate
Elaine A Dunlop et al.
Autophagy, 10(10), 1749-1760 (2014-08-16)
Birt-Hogg-Dubé (BHD) syndrome is a rare autosomal dominant condition caused by mutations in the FLCN gene and characterized by benign hair follicle tumors, pneumothorax, and renal cancer. Folliculin (FLCN), the protein product of the FLCN gene, is a poorly characterized
Katrina Montales et al.
Scientific reports, 11(1), 467-467 (2021-01-13)
The protein kinase ATR is activated at sites of DNA double-strand breaks where it plays important roles in promoting DNA end resection and regulating cell cycle progression. TOPBP1 is a multi BRCT repeat containing protein that activates ATR at DSBs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service