Skip to Content
Merck

06-237

Anti-Gsα Antibody

serum, Upstate®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
17
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, bovine

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GNAS(2778)

General description

large isoform (52 kDa) and small isoform (45 kDa)

Immunogen

peptide corresponding to the c-terminus of bovine Gsα (RMHLRQYELL); this sequence is present in all three isoforms of human Gsα (Gsα1, Gsα2, and GsαXL)

Application

Anti-Gsα Antibody is an antibody against Gsα for use in IC, IP & WB.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Biochem/physiol Actions

Recognizes the large and small forms of Gsα; does not cross-react with other G proteins.

Physical form

Protein A purified
Serum diluted 1:1 in PBS.

Preparation Note

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Widely expressed
routinely evaluated by immunoblot on rat brain microsomal preparation (Catalog #12-144)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB1639

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yan Zhu et al.
Endocrinology, 157(2), 497-507 (2015-12-17)
PTH regulates serum calcium, phosphate, and 1,25-dihydroxyvitamin D (1,25(OH)2D) levels by acting on bone and kidney. In renal proximal tubules (PTs), PTH inhibits reabsorption of phosphate and stimulates the synthesis of 1,25(OH)2D. The PTH receptor couples to multiple G proteins.
Bo Li et al.
FEBS letters, 587(5), 410-416 (2013-01-29)
β-Arrestins function to mediate G protein-coupled receptor (GPCR) desensitization and internalization and to initiate G protein independent signaling of GPCRs. Elucidating how β-arrestin and G protein signal pathways coordinate with each other is important to fully understand GPCR signaling. Here
Arrestin-3 is essential for the activation of Fyn by the luteinizing hormone receptor (LHR) in MA-10 cells.
Colette Galet,Mario Ascoli
Cellular Signalling null



Global Trade Item Number

SKUGTIN
06-23704053252337208