Skip to Content

05-983

Anti-PKC Antibody (α, β, γ), clone M110

clone M110, Upstate®, from mouse

Synonym(s):

PRKCA, PRKCB, PRKCG, protein kinase C, alpha, beta, gamma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
M110, monoclonal
Application:
ELISA, ICC, IP, WB
Citations:
17
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

M110, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PRKCA(5578)

General description

80-82 kDa

Immunogen

Recombinant protein of human PKCγ amino acids 499-697.

Application

Anti-PKC (α Antibody, β, γ), clone M110 detects level of PKC (α & has been published & validated for use in ELISA, IP, WB & IC.

Biochem/physiol Actions

Recognizes PKC α, β, γ. Does not cross react with other PKC isoforms.

Physical form

100 μL of protein A purified mouse monoclonal IgG1 in 50% affinity storage buffer (PBS containing 1.0 mg/mL BSA and 0.05% sodium azide) and 50% glycerol.
Format: Purified

Preparation Note

Stable for 2 years at -20°C from date of shipment. Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Routinely evaluated by immunoblot.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library