05-950
Anti-p54nrb/NonO Antibody, clone 78-1-C6
clone 78-1-C6, Upstate®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
78-1-C6, monoclonal
species reactivity
human, rat
manufacturer/tradename
Upstate®
technique(s)
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... NONO(4841)
Specificity
p54nrb/NonO
Immunogen
KLH-conjugated, synthetic peptide corresponding to amino acids TVREAGPPAFYRPNS
Application
Anti-p54nrb/NonO Antibody, clone 78-1-C6 is a high quality Mouse Monoclonal Antibody for the detection of p54nrb/NonO & has been validated in IP, WB & IHC.
Research Category
Protein Trafficking
Protein Trafficking
Research Sub Category
RNA Metabolism & Binding Proteins
RNA Binding Protein (RBP)
RNA Metabolism & Binding Proteins
RNA Binding Protein (RBP)
Quality
Routinely evaluated by immunoblot.
Target description
54kDa
Physical form
Format: Purified
Protein G Purified
Storage and Stability
2 years at -20°C from date of shipment
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kazumi Nakano et al.
Viruses, 14(2) (2022-02-27)
After integration to the human genome as a provirus, human T-cell leukemia virus type 1 (HTLV-1) utilizes host T cell gene expression machinery for viral replication. The viral RNA-binding protein, Rex, is known to transport unspliced/incompletely spliced viral mRNAs encoding
Xinyu Li et al.
Nucleic acids research, 49(17), 9711-9723 (2021-08-12)
Human fetal globin (γ-globin) genes are developmentally silenced after birth, and reactivation of γ-globin expression in adulthood ameliorates symptoms of hemoglobin disorders, such as sickle cell disease (SCD) and β-thalassemia. However, the mechanisms by which γ-globin expression is precisely regulated
Jenna E Eschbach et al.
mBio, 15(5), e0034824-e0034824 (2024-03-26)
A critical determinant for early post-entry events, the HIV-1 capsid (CA) protein forms the conical core when it rearranges around the dimeric RNA genome and associated viral proteins. Although mutations in CA have been reported to alter innate immune sensing
Yasnee Beeharry et al.
Scientific reports, 8(1), 6031-6031 (2018-04-18)
The Hepatitis Delta Virus (HDV) relies mainly on host proteins for its replication. We previously identified that PSF and p54nrb associate with the HDV RNA genome during viral replication. Together with PSP1, these proteins are part of paraspeckles, which are
Timothy A Vickers et al.
PloS one, 11(8), e0161930-e0161930 (2016-08-30)
Protein-nucleic acid interactions play a crucial role in the regulation of diverse biological processes. Elucidating the roles that protein-nucleic acid complexes play in the regulation of transcription, translation, DNA replication, repair and recombination, and RNA processing continues to be a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service