Skip to Content
Merck
All Photos(1)

Key Documents

05-826

Sigma-Aldrich

Anti-Bcl-2 Antibody, clone AW604

clone AW604, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
AW604, monoclonal
application:
IP
WB
species reactivity:
human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
13

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AW604, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BCL2(596)

Specificity

Bcl-2

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 41-54 (C-GAAPAPGIFSSQPG) of human Bcl-2

Application

Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
This Anti-Bcl-2 Antibody, clone AW604 is validated for use in IP, WB for the detection of Bcl-2.

Quality

routinely evaluated by immunoblot on Raji RIPA cell lysate

Target description

26kDa

Linkage

Replaces: 04-436; OP60-100UG

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Purified

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Programmed cell death: apoptosis and oncogenesis.
Williams, G T
Cell, 65, 1097-1098 (1991)
Ekaterina Kim et al.
Leukemia, 34(6), 1588-1598 (2019-12-22)
Despite major improvements in treatment outcome with novel targeted therapies, such as the Bruton tyrosine kinase (BTK) inhibitor ibrutinib, chronic lymphocytic leukemia (CLL) remains incurable in the majority of patients. Activation of PI3K, NF-κB, and/or MYC has been linked to
S Krajewski et al.
Cancer research, 53(19), 4701-4714 (1993-10-01)
A multidisciplinary approach was taken to investigate the intracellular locations of the 26-kDa integral membrane protein encoded by the bcl-2 gene. Subcellular fractionation analysis of a t(14;18)-containing lymphoma cell line revealed the presence of Bcl-2 protein in nuclear, heavy-membrane, and
Maria Elena Mantione et al.
Cell death & disease, 15(3), 224-224 (2024-03-18)
Microenvironmental signals strongly influence chronic lymphocytic leukemia (CLL) cells through the activation of distinct membrane receptors, such as B-cell receptors, and inflammatory receptors, such as Toll-like receptors (TLRs). Inflammatory pathways downstream of these receptors lead to NF-κB activation, thus protecting
Maria Elena Mantione et al.
Frontiers in oncology, 12, 894413-894413 (2022-07-12)
Renal cell carcinoma is highly inflamed, and tumor cells are embedded into a microenvironment enriched with IL1. While inflammatory pathways are well characterized in the immune system, less is known about these same pathways in epithelial cells; it is unclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service