Skip to Content

05-644

Anti-CAR Antibody, clone RmcB

clone RmcB, Upstate®, from mouse

Synonym(s):

46 kD coxsackievirus and adenovirus receptor (CAR) protein, CVB3 binding protein, CVB3-binding protein, Coxsackievirus B-adenovirus receptor, coxsackie virus B receptor, coxsackie virus and adenovirus receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
RmcB, monoclonal
Application:
ICC, WB
Citations:
56
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RmcB, monoclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CXADR(1525)

General description

The coxsackievirus and adenovirus receptor (CAR) mediates cell attachment and infection by coxsackie B viruses and by a number of adenoviruses. CAR also mediates homotypic intercellular interactions. In polarized epithelial cells, CAR is closely associated with the tight junction, where it contributes to the barrier to paracellular flow of solutes and macromolecules. CAR′s biological roles are not well defined, but emerging evidence suggests that it may function during embryonic development and in regulating cell proliferation.
~46 kDa

Immunogen

Native human coxsackie virus-adenovirus receptor protein.

Application

Anti-CAR Antibody, clone RmcB detects level of CAR & has been published & validated for use in IC & WB.
Immunocytochemistry:
1-2 μg/mL of a previous lot showed positive immunostaining for CAR in HeLa cells, hCAR transfected CHO cells, but not in untransfected CHO cells fixed with 4% paraformaldehyde and permeabilized with 0.2% Triton-X.

Biochem/physiol Actions

Recognizes CAR at MW ~46 kDa.

Physical form

Format: Purified
Purified mouse monoclonal IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl and 0.05% Sodium Azide, pH 7.4.

Analysis Note

Evaluated by Western Blot on mouse small intestine lysates.

Western Blot Analysis:
1:500 dilution of this antibody detected Car on 10 µg of mouse small intestine lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library