Skip to Content
Merck
All Photos(1)

Key Documents

05-414

Sigma-Aldrich

Anti-CrkL Antibody, clone 5-6

clone 5-6, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5-6, monoclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CRKL(1399)

General description

CRKL is a 39kDa adaptor protein with one SH2 and two SH3 binding domains. CRKL is the major protein which is tyrosine phosphorylated in response to BCR/ABL (chronic myelogenous leukemia) and growth factor activation. In myeloid cells, it binds to proteins such as paxillin, p130cas and p120 cbl. CRKL aslo appears to bind ABL and SOS.
The antibody does not cross-react with CRKI or CRKII.

Specificity

Other species have not been tested.
the C-terminal SH3 of Human CRKL

Immunogen

Epitope: Full-length
Full length GST fusion protein corresponding to Human CRKL

Application

Detect CrkL using this Anti-CrkL Antibody, clone 5-6 validated for use in IP, WB, IH.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by Western Blot on K562 cell lysate.

Western Blot Analysis: Antibody detected CRKL in 15 μg of K562 cell lysate.

Target description

39kDa

Physical form

Format: Purified
Protein G Purified
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
Western Blot and Immunoprecipitation:
K562 cell lysate
Immunocytochemistry:
Hela cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eike R Hrincius et al.
Cellular microbiology, 12(6), 831-843 (2010-01-22)
The non-structural protein 1 (A/NS1) of influenza A viruses (IAV) harbours several src-homology domain (SH) binding motifs that are required for interaction with cellular proteins. The SH3 binding motif at aa212-217 [PPLPPK] of A/NS1 was shown to be essential for
Cellular interactions of CRKL, and SH2-SH3 adaptor protein
ten Hoeve, J, et al
Cancer Research, 54, 2563-2567 (1994)
IL-4 regulates the expression of CD209 and subsequent uptake of Mycobacterium leprae by Schwann cells in human leprosy.
Teles RM, Krutzik SR, Ochoa MT, Oliveira RB, Sarno EN, Modlin RL
Infection and Immunity null
The proto-oncogene product p120CBL and the adaptor proteins CRKL and c-CRK link c-ABL, p190BCR/ABL and p210BCR/ABL to the phosphatidylinositol-3' kinase pathway
Sattler, M, et al
Oncogene, 12, 839-846 (1996)
Atsushi Oda et al.
The Journal of biological chemistry, 278(8), 6456-6460 (2003-01-11)
Searching for proteins in platelets that can interact with the N-terminal SH3 domain of CrkL (using a combination of a pull-down assay followed by mass spectrometry), we have found that human platelets express an ADP-ribosylation factor (Arf)-specific GTPase-activating protein (GAP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service