Skip to Content
Merck

05-1946

Anti-USP7 Antibody, clone 1G7

clone 1G7, from mouse

Synonym(s):

ubiquitin specific peptidase 7 (herpes virus-associated), Ubiquitin-specific-processing protease 7, ubiquitin specific protease 7 (herpes virus-associated), Herpesvirus-associated ubiquitin-specific protease, Herpes virus-associated ubiquitin-specific pr

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
1G7, monoclonal
Application:
IHC, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

1G7, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... USP7(7874)

General description

Ubiquitin carboxyl-terminal hydrolase 7 (USP7) is a ubiquitin specific protease or a deubiquitylating enzyme that cleaves ubiquitin from its substrates.[Saridakis, 2005] Since ubiquitylation (polyubiquitination) is most commonly associated with the stability and degradation of cellular proteins, Ubiquitin carboxyl-terminal hydrolase 7 activity generally stabilizes its substrate proteins. Ubiquitin carboxyl-terminal hydrolase 7 is most popularly known as a direct antagonist of Mdm2, the E3 ubiquitin ligase for the tumor suppressor protein, p53.[Canning, 2004]
~ 128kDa Observed

Immunogen

Histadine-tagged recombinant protein corresponding to human USP7.

Application

This Anti-USP7 Antibody, clone 1G7 is validated for use in WB, IP, IH for the detection of USP7.

Biochem/physiol Actions

This antibody recognizes USP7.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blot in HeLa lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected USP7 on 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
05-194604053252628818