Skip to Content
Merck
All Photos(1)

Key Documents

05-1517

Sigma-Aldrich

Anti-hnRNP Q Antibody, clone 18E4

clone 18E4, from mouse

Synonym(s):

heterogeneous nuclear ribonucleoprotein Q-like, Synaptotagmin-binding, cytoplasmic RNA-interacting protein, Glycine- and tyrosine-rich RNA-binding protein, GRY-RBP, NS1-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
18E4, monoclonal
application:
ICC
IP
WB
species reactivity:
human
technique(s):
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
6

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

18E4, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYNCRIP(10492)

General description

Heterogeneous nuclear ribonucleoprotein Q commoly refered to as hnRNP Q can be found as three different isoforms derived from alternative splicing of a single gene. This RNA binding protein is implicated in control of mRNA precursor splicing for the survival of motor neurons (SMN) protein, a gene. Loss-of-function mutations of SMN are linked to spinal muscular atrophy (SMA) a common neurodegenerative disease. It has been shown that hnRNP Q proteins interact with SMN, and this is interacton is required for efficient pre-mRNA splicing in vitro. Current data suggests that hnRNP Q is a splicing modulator of SMN.

Specificity

This antibody recognizes hnRNP Q.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human hnRNP Q.

Application

Immunocytochemistry
Cited by independent researcher using a representative lot.
Immunoprecipitation
Cited by independent researcher using a representative lot.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-hnRNP Q Antibody, clone 18E4 (Mouse Monoclonal Antibody) validated in WB, ICC, IP to detect hnRNP Q also known as heterogeneous nuclear ribonucleoprotein Q-like.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/ml of this antibody detected hnRNP Q on 10 µg of HeLa cell lysate.

Target description

The calculated molecular weight is 70 kDa Clone 18E4 can recognizes four bands on the Western Blot of total HeLa cell lysate corresponding to molecular masses of ~80, ~70, ~60 and ~55 kDa hnRNP Q has three isoforms of ~70– 55 kDa The ~80 kDa reactive band was identified as hnRNP R, which migrates at ~80 kDa on SDS–PAGE. (Mourelatos., Z., et al., EMBO J., 20, 5443-5452 (2001).)

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgGκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuri V Svitkin et al.
PLoS biology, 11(5), e1001564-e1001564 (2013-05-24)
Translation control often operates via remodeling of messenger ribonucleoprotein particles. The poly(A) binding protein (PABP) simultaneously interacts with the 3' poly(A) tail of the mRNA and the eukaryotic translation initiation factor 4G (eIF4G) to stimulate translation. PABP also promotes miRNA-dependent
Fruzsina Hobor et al.
Nature communications, 9(1), 831-831 (2018-02-28)
Exosomal miRNA transfer is a mechanism for cell-cell communication that is important in the immune response, in the functioning of the nervous system and in cancer. Syncrip/hnRNPQ is a highly conserved RNA-binding protein that mediates the exosomal partition of a
Ruiping Zheng et al.
Journal of cell science, 123(Pt 21), 3734-3744 (2010-10-14)
In higher eukaryotic cells, long non-protein-coding RNAs (lncRNAs) have been implicated in a wide array of cellular functions. Cell- or tissue-specific expression of lncRNA genes encoded in the mammalian genome is thought to contribute to the complex gene networks needed
Lauren M Gittings et al.
Acta neuropathologica communications, 7(1), 18-18 (2019-02-14)
Frontotemporal lobar degeneration (FTLD) is pathologically subdivided based on the presence of particular pathological proteins that are identified in inclusion bodies observed post-mortem. The FTLD-FUS subgroup is defined by the presence of the fused in sarcoma protein (FUS) in pathological
Xiangling Meng et al.
Nature, 622(7982), 359-366 (2023-09-28)
The assembly of cortical circuits involves the generation and migration of interneurons from the ventral to the dorsal forebrain1-3, which has been challenging to study at inaccessible stages of late gestation and early postnatal human development4. Autism spectrum disorder and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service