Skip to Content
Merck
All Photos(1)

Key Documents

05-1243

Sigma-Aldrich

Anti-Sirt1 Antibody, clone 3H10.2

clone 3H10.2, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H10.2, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRT1(23411)

General description

The Sir2 protein in yeast is known to function in transcriptional silencing processes through the deacetylation of histones H3 and H4. The more recently described human homologue of Sir2, known as SIRT1, has been found to associate with the tumor suppressor protein p53. There are now five described human homologs of yeast Sir2, which are named SIRT1-5.
SIRT1 binds and deacetylates p53 with specificity for its C-terminal Lys382 residue in response to the upregulation of promyelocytic leukemia protein (PML) nuclear bodies or oncogenic Ras. The deacetylation of p53 SIRT1 has been shown to negatively regulate p53-mediated transcription, preventing cellular senescence and apoptosis induced by DNA damage and stress. SIRT1 is a HDAC which is important in establishing repressive chromatin structures and plays a role in increasing lifespan.

Specificity

Human Sirt1. Does not cross react with lysates from mouse NIH/3T3 cells or rat PC12 cells.

Immunogen

GST fusion protein corresponding to amino acids 508-616 of human Sirt1.

Application

Anti-Sirt1 Antibody, clone 3H10.2 is a Mouse Monoclonal Antibody for detection of Sirt1 also known as Sirtuin 1, Silent mating type Information Regulation-2 & has been tested in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot on nuclear extract from HeLa cells

Target description

120 kDa

Linkage

Replaces: 05-707

Physical form

Format: Purified
Protein G Purified
Purified in PBS with 0.05% Sodium Azide

Storage and Stability

Maintain at 2-8 °C in undiluted aliquots for up to 6 months after date of receipt. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Coutelle et al.
EMBO molecular medicine, 6(5), 624-639 (2014-03-22)
In the normal quiescent vasculature, only 0.01% of endothelial cells (ECs) are proliferating. However, this proportion increases dramatically following the angiogenic switch during tumor growth or wound healing. Recent evidence suggests that this angiogenic switch is accompanied by a metabolic
p53 Activation: a case against Sir.
Brooks, Christopher L and Gu, Wei
Cancer Cell, 13, 377-378 (2008)
Hsiang-Yu Chang et al.
Nature communications, 4, 2757-2757 (2013-11-14)
Conformational disorders are involved in various neurodegenerative diseases. Reactive oxygen species (ROS) are the major contributors to neurodegenerative disease; however, ROS that affect the structural changes in misfolded disease proteins have yet to be well characterized. Here we demonstrate that
Timour Prozorovski et al.
Nature cell biology, 10(4), 385-394 (2008-03-18)
Repair processes that are activated in response to neuronal injury, be it inflammatory, ischaemic, metabolic, traumatic or other cause, are characterized by a failure to replenish neurons and by astrogliosis. The underlying molecular pathways, however, are poorly understood. Here, we
Peter J Elliott et al.
Current opinion in investigational drugs (London, England : 2000), 9(4), 371-378 (2008-04-09)
Sirtuins represent a novel family of enzymes that are collectively well situated to help regulate nutrient sensing and utilization, metabolic rate and ultimately metabolic disease. Activation of one of these enzymes, SIRT1, leads to enhanced activity of multiple proteins, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service