Skip to Content
Merck
All Photos(2)

Key Documents

04-928

Sigma-Aldrich

Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal

culture supernatant, clone A3S, Upstate®

Synonym(s):

H3, Histone H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
A3S, monoclonal
application:
ChIP
WB
species reactivity:
human, rat, Saccharomyces cerevisiae, mouse, chicken
technique(s):
ChIP: suitable
western blot: suitable
citations:
23

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

A3S, monoclonal

species reactivity

human, rat, Saccharomyces cerevisiae, mouse, chicken

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... H3F3B(3021)

General description

An antibody that recognizes the C-terminus of histone H3 and is useful for identifying total H3 independant of post-translational modifications.

Specificity

Broad species cross-reactivity expected due to sequence homology
Recognizes Histone H3.

Immunogen

KLH-conjugated, synthetic peptide corresponding to the C-terminus of human Histone H3

Application

Anti-Histone H3 Antibody, CT, pan, clone A3S is a Rabbit Monoclonal Antibody for detection of Histone H3 also known as Histone H3 & has been validated in ChIP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot Analysis: 1:500-1:2,000 dilution of this lot detected Histone H3 in a modification independent manner in acid extracted proteins from untreated, sodium butyrate or colcemid treated HeLa cells.

Chromatin Immunoprecipitation:
2 μL of of a previous lot of this antibody immunoprecipitated chromatin associated with Histone H3 from a wild type yeast lysate.

Quality

Routinely evaluated by immunoblot analysis.

Target description

17 kDa

Linkage

Replaces: 05-928

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Kain et al.
Molecular metabolism, 53, 101291-101291 (2021-07-12)
Type II nuclear hormone receptors, including farnesoid X receptors (FXR), liver X receptors (LXR), and peroxisome proliferator-activated receptors (PPAR), which serve as drug targets for metabolic diseases, are permanently positioned in the nucleus and thought to be bound to DNA
Huiyuan Zhang et al.
Nature communications, 5, 5798-5798 (2014-12-17)
The transcriptional regulator STAT3 curbs pro-inflammatory cytokine production mediated by NF-κB signalling in innate immune cells, yet the mechanism by which this occurs has been unclear. Here we identify STAT3 as a pivotal negative regulator of Ubc13, an E2 ubiquitin-conjugating
Marcel Donnet et al.
Journal of colloid and interface science, 340(2), 218-224 (2009-10-10)
Thermodynamic solubility calculations are normally only related to thermodynamic equilibria in solution. In this paper, we extend the use of such solubility calculations to help elucidate possible precipitation reaction pathways during the entire reaction. We also estimate the interfacial energy
Histone H3 lysine 4 methylation is mediated by Set1 and required for cell growth and rDNA silencing in Saccharomyces cerevisiae.
Briggs, S D, et al.
Genes & Development, 15, 3286-3295 (2001)
Michelle L Larin et al.
PLoS genetics, 11(11), e1005425-e1005425 (2015-11-21)
Changes in the locations and boundaries of heterochromatin are critical during development, and de novo assembly of silent chromatin in budding yeast is a well-studied model for how new sites of heterochromatin assemble. De novo assembly cannot occur in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service