Skip to Content
Merck
All Photos(1)

Key Documents

04-469

Sigma-Aldrich

Anti-SREBP1 Antibody, clone 2121

clone 2121, Upstate®, from mouse

Synonym(s):

SREBF1, sterol regulatory element binding transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2121, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SREBF1(6720)

Specificity

Reacts with the basic helix-loop-helix leucine zipper (bHLH-ZIP) of SREBP.

Immunogen

Recombinant His-tagged human SREBP-1a protein encompassing amino acids 301-407.

Application

Anti-SREBP1 Antibody, clone 2121 is a Mouse Monoclonal Antibody for detection of SREBP1 also known as sterol regulatory element binding transcription factor 1 & has been validated in WB.

Quality

Routinely evaluated by immunoblot.

Target description

126 kDa

Physical form

100 μL (1 mg/mL) of protein A purified mouse monoclonal IgG1 in PBS with 0.1% sodium azide.
Format: Purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment. Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Other Notes

Please refer to lot specific datasheet.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanel Punga et al.
The Journal of biological chemistry, 281(35), 25278-25286 (2006-07-11)
Members of the sterol regulatory element-binding protein (SREBP) family of transcription factors control cholesterol and lipid metabolism and play critical roles during adipocyte differentiation. The transcription factor SREBP1 is degraded by the ubiquitin-proteasome system following phosphorylation of Thr426 and Ser430
Shreetama Bandyopadhayaya et al.
Journal of biochemical and molecular toxicology, 35(4), e22693-e22693 (2021-01-05)
N-acyl dopamines (NADAs) are bioactive lipids of the endovanilloid family with known cytotoxicity for the cancer cells; however, the available data on the participation of the endovanilloids in epithelial-mesenchymal transition (EMT) and cancer stemness are controversial. This study unveils the
X Wang et al.
The Journal of biological chemistry, 268(19), 14497-14504 (1993-07-05)
This paper describes the purification and characterization of a sterol regulatory element binding protein (SREBP) that recognizes the SRE-1 sequence in the 5' flanking region of the gene for the low density lipoprotein (LDL) receptor. The protein was purified more
Amy K Walker et al.
Genes & development, 24(13), 1403-1417 (2010-07-03)
The sterol regulatory element-binding protein (SREBP) transcription factor family is a critical regulator of lipid and sterol homeostasis in eukaryotes. In mammals, SREBPs are highly active in the fed state to promote the expression of lipogenic and cholesterogenic genes and
Yuichi Wakana et al.
The Journal of cell biology, 220(1) (2020-11-07)
In response to cholesterol deprivation, SCAP escorts SREBP transcription factors from the endoplasmic reticulum to the Golgi complex for their proteolytic activation, leading to gene expression for cholesterol synthesis and uptake. Here, we show that in cholesterol-fed cells, ER-localized SCAP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service